Learn More
Medchemexpress LLC HDAC10-IN-2 hydrochloride | 00-00-0 | ≥95.0% | 346.85 g/mol | C19H23ClN2O2 | 5 MG
SDP
Catalog No. 5000221660
Encompass_Preferred
HDAC10-IN-2 hydrochloride is a research-grade, selective inhibitor of histone deacetylase 10 (HDAC10). It modulates autophagy in FLT3-ITD-positive acute myeloid leukemia cells and is supplied as the hydrochloride salt with high purity for laboratory use.
  • Selective HDAC10 inhibitor with an IC50 of 20 nM.
  • High purity (≥95.0%) suitable for biochemical and cellular assays.
  • Supplied as a hydrochloride salt to improve stability and solubility.
  • Available in small research quantities (5 mg and larger packaging sizes).
  • Store sealed at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-21660 Supplier Medchemexpress LLC Supplier No. HY144782A5MG
HDAC10-IN-2 hydrochloride is a research-grade, selective inhibitor of histone deacetylase 10 (HDAC10). It modulates autophagy in FLT3-ITD-positive acute myeloid leukemia cells and is supplied as the hydrochloride salt with high purity for laboratory use.
  • Selective HDAC10 inhibitor with an IC50 of 20 nM.
  • High purity (≥95.0%) suitable for biochemical and cellular assays.
  • Supplied as a hydrochloride salt to improve stability and solubility.
  • Available in small research quantities (5 mg and larger packaging sizes).
  • Store sealed at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month).