Learn More
Medchemexpress LLC HDAC10-IN-2 hydrochloride | 00-00-0 | 95.0% | C19H23ClN2O2 | 10MG
SDP
Catalog No. 5000258516
Encompass_Preferred
HDAC10-IN-2 hydrochloride is a potent, selective histone deacetylase 10 (HDAC10) inhibitor for research use that modulates autophagy in FLT3-ITD-positive acute myeloid leukemia cells. It exhibits low-nanomolar potency and is supplied as a solid hydrochloride salt with recommended storage conditions.
  • Potent HDAC10 inhibition (IC50 ≈ 20 ± 2 nM)
  • Selective over HDAC8 and HDAC6 (reported selectivity data)
  • Modulates autophagy in FLT3-ITD-positive acute myeloid leukemia cells
  • Reported purity 95.0%
  • Supplied in multiple small-scale sizes suitable for research
  • Store sealed, away from moisture and light; in solution, -80°C (6 months) or -20°C (1 month)
Catalog No. 50-002-58516 Supplier Medchemexpress LLC Supplier No. HY144782A10MG
HDAC10-IN-2 hydrochloride is a potent, selective histone deacetylase 10 (HDAC10) inhibitor for research use that modulates autophagy in FLT3-ITD-positive acute myeloid leukemia cells. It exhibits low-nanomolar potency and is supplied as a solid hydrochloride salt with recommended storage conditions.
  • Potent HDAC10 inhibition (IC50 ≈ 20 ± 2 nM)
  • Selective over HDAC8 and HDAC6 (reported selectivity data)
  • Modulates autophagy in FLT3-ITD-positive acute myeloid leukemia cells
  • Reported purity 95.0%
  • Supplied in multiple small-scale sizes suitable for research
  • Store sealed, away from moisture and light; in solution, -80°C (6 months) or -20°C (1 month)