Learn More
Medchemexpress LLC Hdac10-in-2 hydrochloride | 95.0% | C19H23ClN2O2 | 100 MG
SDP
Catalog No. 5000356677
Encompass_Preferred
HDAC10-IN-2 hydrochloride is a potent and highly selective HDAC10 inhibitor with an IC50 of 20 nM. It modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells. This compound is for research use only and appears as an off-white to yellow solid with a molecular weight of 346.85.
  • Potent and highly selective HDAC10 inhibitor
  • Modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells
  • Appears as an off-white to yellow solid
  • For research use only
Catalog No. 50-003-56677 Supplier Medchemexpress LLC Supplier No. HY144782A100MG
HDAC10-IN-2 hydrochloride is a potent and highly selective HDAC10 inhibitor with an IC50 of 20 nM. It modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells. This compound is for research use only and appears as an off-white to yellow solid with a molecular weight of 346.85.
  • Potent and highly selective HDAC10 inhibitor
  • Modulates autophagy in aggressive FLT3-ITD positive acute myeloid leukemia cells
  • Appears as an off-white to yellow solid
  • For research use only