Learn More
ABclonal Technology HDAC11 Rabbit mAb
SDP
Catalog No. 5000012241
Encompass_Preferred
This gene encodes a class IV histone deacetylase The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10 Alternative splicing results in multiple transcript variants
Catalog No. 50-000-12241 Supplier ABclonal Technology Supplier No. A25809100UL
This gene encodes a class IV histone deacetylase The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10 Alternative splicing results in multiple transcript variants