Learn More
Aviva Systems Biology Corp HDAC2 Antibody - middle region (ARP32619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HDAC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HDAC2. Alternate names: RPD3, YAF1, HD2ApplicationCHIP, IHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63082