Learn More
Enzo Life Sciences FLUOR DE LYS-Green HDAC2 fluorometric drug discovery assay kit (1x96wells)
SDP
Catalog No. 502010286
Encompass_Preferred
The HDAC2 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human HDAC2 included in the kit. The kit is ideal for chemical library screening for candidate inhibitors or kinetic assay of the enzyme under varying conditions. The FLUOR DE LYS-Green HDAC2 assay is based on the FLUOR DE LYS-Green Substrate and FLUOR DE LYS Developer combination. The assay procedure has two steps. First, the FLUOR DE LYS-Green Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC2. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYS Developer produces a fluorophore. Alternative name: Histone deacetylase 2 fluorescent assay kit. Applications: Fluorescent detection, HTS Cell-based assays, Activity assay.
Catalog No. 50-201-0286 Supplier Enzo Life Sciences Supplier No. BMLAK5120001
The HDAC2 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human HDAC2 included in the kit. The kit is ideal for chemical library screening for candidate inhibitors or kinetic assay of the enzyme under varying conditions. The FLUOR DE LYS-Green HDAC2 assay is based on the FLUOR DE LYS-Green Substrate and FLUOR DE LYS Developer combination. The assay procedure has two steps. First, the FLUOR DE LYS-Green Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC2. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYS Developer produces a fluorophore. Alternative name: Histone deacetylase 2 fluorescent assay kit. Applications: Fluorescent detection, HTS Cell-based assays, Activity assay.