Learn More
Enzo Life Sciences HDAC2 (full-length) (human), (recombinant) (His-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5330050
Encompass_Preferred
Alternative name: Histone deacetylase 2. Formulation: Liquid. In 50mM TRIS, pH 8.0, 138 mM NaCl and 10% glycerol. Source: Produced in insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2072