Learn More
Enzo Life Sciences HDAC2 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA4020100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 475-488 of C-terminal human HDAC2. Species reactivity: Human, Mouse, Rat Hamster, Dog. Applications: ICC, IHC, IP, WB. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-1812