Learn More
Aviva Systems Biology Corp Hdac3 Antibody - C-terminal region (ARP42786_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42786_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hdac3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Hdac3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-025