Learn More
Enzo Life Sciences HDAC3 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA4030100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 415-428 of C-terminal human HDAC3. Species reactivity: Human, Rat Hamster, Dog. Applications: WB. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-1813