Learn More
Sino Biological Recombinant Human RhoA Protein (Mature form, His Tag), Active
Catalog No. 502573247
Encompass_Preferred
Recombinant human RhoA (1-190) was expressed in E.coli cells using an N-terminal His tag.
Catalog No. 50-257-3247 Supplier Sino Biological Supplier No. R144310H50
Recombinant human RhoA (1-190) was expressed in E.coli cells using an N-terminal His tag.