Learn More
Aviva Systems Biology Corp HDAC4 Antibody - N-terminal region (ARP34229_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34229_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HDAC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HDAC4. Alternate names: HA6116, HD4, HDAC-A, HDACA, KIAA0288, AHO3, BDMR, HDAC-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55309