Learn More
Enzo Life Sciences HDAC5 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210340C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 572-587 of human HDAC5 (histone deacetylase 5). Species reactivity: Human, Mouse. Applications: IP, WB ChIP. Purity: Protein G-affinity purified IgG. Formulation: Liquid. In PBS containing 0.05% sodium azide. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8115