Learn More
Aviva Systems Biology Corp Hdac6 Antibody - C-terminal region (ARP38730_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hdac6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse Hdac6. Alternate names: Hd6, Hdac5, Sfc6, mHDA2ApplicationWB, CHIP, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-757