Learn More
Medchemexpress LLC HDAC6 degrader-1 | 2439058-23-6 | 95.7% | 795.84 | C40H45N9O9 | 5 MG
SDP
Catalog No. 5000262273
Encompass_Preferred
HDAC6 degrader-1 is a proteolysis-targeting chimera (PROTAC) designed to selectively induce degradation of histone deacetylase 6 (HDAC6) in cell-based studies. It links an HDAC6-binding ligand to an E3 ligase recruiter to promote ubiquitination and proteasomal clearance of the target protein, and has been reported to show potent cellular activity.
  • Selective degradation of HDAC6 in cells
  • Potent cellular activity: DC50 3.8 nM in MM.1S cells
  • Research-grade solid with high purity (95.7%)
  • Molecular weight 795.84 for assay planning
  • Available in small research pack sizes suitable for in vitro studies
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month)
Catalog No. 50-002-62273 Supplier Medchemexpress LLC Supplier No. HY1329985MG
HDAC6 degrader-1 is a proteolysis-targeting chimera (PROTAC) designed to selectively induce degradation of histone deacetylase 6 (HDAC6) in cell-based studies. It links an HDAC6-binding ligand to an E3 ligase recruiter to promote ubiquitination and proteasomal clearance of the target protein, and has been reported to show potent cellular activity.
  • Selective degradation of HDAC6 in cells
  • Potent cellular activity: DC50 3.8 nM in MM.1S cells
  • Research-grade solid with high purity (95.7%)
  • Molecular weight 795.84 for assay planning
  • Available in small research pack sizes suitable for in vitro studies
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month)