Learn More
Medchemexpress LLC HDAC6-IN-33 | 2444302-23-0 | 99.0% | 303.27 | 1 ML
SDP
Catalog No. 5000394544
Encompass_Preferred
HDAC6-IN-33 (compound 6) is a selective and irreversible HDAC6 inhibitor with an IC50 of 193 nM. It demonstrates no activity against HDAC1-4. This compound is a tight-binding HDAC6 inhibitor that can inhibit HDAC6 through a two-step slow-binding mechanism. It is intended for research use only and is not sold to patients.
  • Selective and irreversible HDAC6 inhibitor.
  • IC50 of 193 nM for HDAC6.
  • Shows no activity against HDAC1-4.
  • Inhibits HDAC6 via a two-step slow-binding mechanism.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg) for purchase.
  • Free samples (0.1 - 0.2 mg) are available for application.
  • Solution in DMSO is available (10 mM * 1 mL).
  • Detailed data sheet, COA, and SDS available.
  • Information on solubility and storage provided.
  • Cited in scientific publications.
Catalog No. 50-003-94544 Supplier Medchemexpress LLC Supplier No. HY16130410MM/1ML
HDAC6-IN-33 (compound 6) is a selective and irreversible HDAC6 inhibitor with an IC50 of 193 nM. It demonstrates no activity against HDAC1-4. This compound is a tight-binding HDAC6 inhibitor that can inhibit HDAC6 through a two-step slow-binding mechanism. It is intended for research use only and is not sold to patients.
  • Selective and irreversible HDAC6 inhibitor.
  • IC50 of 193 nM for HDAC6.
  • Shows no activity against HDAC1-4.
  • Inhibits HDAC6 via a two-step slow-binding mechanism.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg) for purchase.
  • Free samples (0.1 - 0.2 mg) are available for application.
  • Solution in DMSO is available (10 mM * 1 mL).
  • Detailed data sheet, COA, and SDS available.
  • Information on solubility and storage provided.
  • Cited in scientific publications.