Learn More
Cayman Chemical HDAC6 Inhibitor, Purity: ≥95%, CAS: 1259296-46-2, 10 mg
SDP
Catalog No. 502970735
Encompass_Preferred
A potent and selective inhibitor of HDAC6 (IC50 = 36 nM) that poorly blocks other HDAC enzymes; cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM
Catalog No. 50-297-0735 Supplier Cayman Chemical Supplier No. 1520010 MG
A potent and selective inhibitor of HDAC6 (IC50 = 36 nM) that poorly blocks other HDAC enzymes; cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM