Learn More
Cayman Chemical HDAC6 Inhibitor, Purity: ≥95%, CAS: 1259296-46-2, 5 mg
SDP
Catalog No. 502968933
Encompass_Preferred
A potent and selective inhibitor of HDAC6 (IC50 = 36 nM) that poorly blocks other HDAC enzymes; cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM
Catalog No. 50-296-8933 Supplier Cayman Chemical Supplier No. 152005 MG
A potent and selective inhibitor of HDAC6 (IC50 = 36 nM) that poorly blocks other HDAC enzymes; cell permeable, inhibiting the acetylation of tubulin in cells with an IC50 value of 210 nM