Learn More
Aviva Systems Biology Corp HDAC8 Antibody - C-terminal region (ARP34587_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34587_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HDAC8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HDAC8. Alternate names: HDACL1, RPD3, HD8, CDA07ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55465