Learn More
Cayman Chemical HDAC8 Developer 1ea
SDP
Catalog No. 502896769
Encompass_Preferred
A PARP1 and PARP2 inhibitor (IC50s 50s 1-14.1 nM) inhibits the proliferation of Capan-1 cells (IC50 0.1 nM) as well as talazoparib- or olaparib-resistant Capan-1 cells (IC50s 1.13 and 0.89 nM respectively) inhibits the proliferation of BRCA1 mutant HCC1937 breast cancer cells (IC50 4.54 nM) reduces tumor growth in an olaparib-resistant MDA-MB-436 breast cancer mouse xenograft model at 12.5 and 25 mg/kg
Catalog No. 50-289-6769 Supplier Cayman Chemical Supplier No. 7002351 EA
A PARP1 and PARP2 inhibitor (IC50s 50s 1-14.1 nM) inhibits the proliferation of Capan-1 cells (IC50 0.1 nM) as well as talazoparib- or olaparib-resistant Capan-1 cells (IC50s 1.13 and 0.89 nM respectively) inhibits the proliferation of BRCA1 mutant HCC1937 breast cancer cells (IC50 4.54 nM) reduces tumor growth in an olaparib-resistant MDA-MB-436 breast cancer mouse xenograft model at 12.5 and 25 mg/kg