Learn More
Cayman Chemical HDAC8 Fluorophore 1ea
SDP
Catalog No. 502896637
Encompass_Preferred
A negative modulator of mitochondrial calcium uptake selectively decreases mitochondrial over cytosolic histamine-induced calcium uptake but also decreases the mitochondrial membrane potential in HeLa cells at 10 UM binds to Micu1 in a cell-free assay (Kd 8.4 UM for the mouse protein) decreases myotube width in differentiated C2C12 skeletal muscle myoblasts in an in vitro model of myotube growth protects against ferroptosis induced by RSL3 erastin or glutamate in HT22 hippocampal cells at 2.5 UM
Catalog No. 50-289-6637 Supplier Cayman Chemical Supplier No. 7002361 EA
A negative modulator of mitochondrial calcium uptake selectively decreases mitochondrial over cytosolic histamine-induced calcium uptake but also decreases the mitochondrial membrane potential in HeLa cells at 10 UM binds to Micu1 in a cell-free assay (Kd 8.4 UM for the mouse protein) decreases myotube width in differentiated C2C12 skeletal muscle myoblasts in an in vitro model of myotube growth protects against ferroptosis induced by RSL3 erastin or glutamate in HT22 hippocampal cells at 2.5 UM