Learn More
Medchemexpress LLC HY-111342 5mg Medchemexpress, HDAC8-IN-1 CAS:1417997-93-3 Purity:>98%
SDP
Catalog No. 501968834
Encompass_Preferred
Medchemexpress, HY-111342 5mg HDAC8-IN-1 CAS:1417997-93-3 HDAC8-IN-1 is a HDAC8 inhibitor with an IC50 of 27.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8834 Supplier Medchemexpress LLC Supplier No. HY1113425MG
Medchemexpress, HY-111342 5mg HDAC8-IN-1 CAS:1417997-93-3 HDAC8-IN-1 is a HDAC8 inhibitor with an IC50 of 27.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.