Learn More
Aviva Systems Biology Corp HDDC3 Antibody - middle region (ARP55898_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55898_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HDDC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HDDC3. Alternate names: MGC45386, MESH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61655