Learn More
Med Vet International HDE Evolution Dental Set Compact Water-Cooling Level 2, J1410S2CWC, Each
Catalog No. 5000584095
Encompass_Preferred
The Set Level 2 is composed of: • Evolution engine (complete set) with jacket – Watercooling • Quickflex drive shaft 1.20 m – Watercooling • COMPACT Flexxidisc long curved twist – Watercooling • Stainless steel brush with holder • Trolley instrument case • Base plate for handpieces
Catalog No. 50-005-84095 Supplier Med Vet International Supplier No. JOR1410S2CWC
The Set Level 2 is composed of: • Evolution engine (complete set) with jacket – Watercooling • Quickflex drive shaft 1.20 m – Watercooling • COMPACT Flexxidisc long curved twist – Watercooling • Stainless steel brush with holder • Trolley instrument case • Base plate for handpieces