Learn More
Med Vet International HDE Evolution Dental Set Compact Water-Cooling Level 4, J1410S4CWC, Each
Catalog No. 5000584245
Encompass_Preferred
The Set Level 4 is composed of: • Evolution engine (complete set) with jacket – Watercooling • Quickflex drive shaft 1.20 m – Watercooling • COMPACT Flexxidisc long curved twist – Watercooling • COMPACT Polyfloat long straight (without burr) – Watercooling • COMPACT – Applecore Ø14 • COMPACT – Occlusal burr • COMPACT – Diastema long versatile burr • COMPACT – Conical burr long • Handpiece for the 11s – Straight long with closed head protection • Stainless steel brush with holder • Trolley instrument case • Base plate for handpieces (in option)
Catalog No. 50-005-84245 Supplier Med Vet International Supplier No. JOR1410S4CWC
The Set Level 4 is composed of: • Evolution engine (complete set) with jacket – Watercooling • Quickflex drive shaft 1.20 m – Watercooling • COMPACT Flexxidisc long curved twist – Watercooling • COMPACT Polyfloat long straight (without burr) – Watercooling • COMPACT – Applecore Ø14 • COMPACT – Occlusal burr • COMPACT – Diastema long versatile burr • COMPACT – Conical burr long • Handpiece for the 11s – Straight long with closed head protection • Stainless steel brush with holder • Trolley instrument case • Base plate for handpieces (in option)