Learn More
Med Vet International HDE HydroJet II Yellow Kit - Complete Kit, J1417, Each
Supplier:  Med Vet International JOR1417
Encompass_Preferred
Hydro Jet II. Compact and portable, the HydroJet II is the essential tool for proper irrigation, rinsing of debris and efficiently flushing of diastemas as an integral part of equine dentistry. • Rechargeable 14.4a Lithium Battery J1417 Hydro Jet II Set includes: • Hydro Jet II with battery and charger • Rinsing nozzle • Diastema nozzle • 0.26 gal/1-liter bottle • 29.5 ft/6-meter water tube • Slim Carry Case
Catalog No. 50-005-85817