Learn More
Med Vet International HDE SwissFloat SlimLine Dental Float AC Power, J1645SAC, Each
Catalog No. 5000585670
Encompass_Preferred
HDE SwissFloat Slimline Dental Float. Made for easy reach, especially the 11s due to dental float thin head profile. • Easy to change disc by yourself • Float shaft orientation can be changed (mandibular or maxillary arcade). J1645sac HDE SwissFloat Dental Float – AC
Catalog No. 50-005-85670 Supplier Med Vet International Supplier No. JOR1645SAC
HDE SwissFloat Slimline Dental Float. Made for easy reach, especially the 11s due to dental float thin head profile. • Easy to change disc by yourself • Float shaft orientation can be changed (mandibular or maxillary arcade). J1645sac HDE SwissFloat Dental Float – AC