Learn More
Med Vet International HDE Swissfloat Slimline Dental Float – Battery Powered, J1645SB, Each
Catalog No. 5000585747
Encompass_Preferred
HDE SwissFloat Slimline Dental Float. Made for easy reach, especially the 11s due to dental float thin head profile. • Easy to change disc by yourself • Float shaft orientation can be changed (mandibular or maxillary arcade) Cordless
Catalog No. 50-005-85747 Supplier Med Vet International Supplier No. JOR1645SB
HDE SwissFloat Slimline Dental Float. Made for easy reach, especially the 11s due to dental float thin head profile. • Easy to change disc by yourself • Float shaft orientation can be changed (mandibular or maxillary arcade) Cordless