Learn More
Medchemexpress LLC Hdhodh-in-1 | 1173715-42-8 | MFCD32671346 | >98.0% | C17H14N2O2 | 10MG
SDP
Catalog No. 5000258232
Encompass_Preferred
hDHODH-IN-1 is a small-molecule inhibitor of human dihydroorotate dehydrogenase (hDHODH) used in research to probe DHODH-dependent pyrimidine biosynthesis and inflammation-related pathways. It is supplied as a research reagent and has documented activity in biochemical and cellular assays.
  • Inhibits human dihydroorotate dehydrogenase.
  • Useful for studying pyrimidine biosynthesis and inflammatory responses.
  • Applicable in biochemical and cellular assay formats.
  • High purity confirmed by LCMS (>99.9%).
  • Molecular formula C17H14N2O2 and molecular weight 278.31 g/mol.
Catalog No. 50-002-58232 Supplier Medchemexpress LLC Supplier No. HY13565810MG
hDHODH-IN-1 is a small-molecule inhibitor of human dihydroorotate dehydrogenase (hDHODH) used in research to probe DHODH-dependent pyrimidine biosynthesis and inflammation-related pathways. It is supplied as a research reagent and has documented activity in biochemical and cellular assays.
  • Inhibits human dihydroorotate dehydrogenase.
  • Useful for studying pyrimidine biosynthesis and inflammatory responses.
  • Applicable in biochemical and cellular assay formats.
  • High purity confirmed by LCMS (>99.9%).
  • Molecular formula C17H14N2O2 and molecular weight 278.31 g/mol.