Learn More
TARGETMOL CHEMICALS INC HDHODH-IN-7 5MG
SDP
Catalog No. 502581588
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hDHODH-IN-7 (DHODH-IN-9) is a nitrogen-containing analog and is a human dihydrorotatory dehydrogenase inhibitor. hDHODH-IN-7 and pMIC50 of 7.4 have antiviral effects. purity: 99%
Catalog No. 50-258-1588 Supplier TARGETMOL CHEMICALS INC Supplier No. T110315MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hDHODH-IN-7 (DHODH-IN-9) is a nitrogen-containing analog and is a human dihydrorotatory dehydrogenase inhibitor. hDHODH-IN-7 and pMIC50 of 7.4 have antiviral effects. purity: 99%