Learn More
MilliporeSigma HDL
Supplier:  MilliporeSigma LP35MG
Encompass
HDL; Chemicon International; Purity: > 98%; 5mg; M.W.: 175000-360000; Density: 1.063-1.2g/mL; Liquid in 150mM NaCl; Prepared by sequential flotation ultracentrifugation technique; Storage: 2 deg. to 8 deg.C upto 4 weeks
Catalog No. 50-571-24