Learn More
Cell Biolabs Inc HDL and LDL/VLDL Cholesterol Assay Kit, 192 assays
Catalog No. 50673578
Encompass_Preferred
Cell Biolabs’ HDL and LDL/VLDL Cholesterol Assay Kit is a simple fluorometric assay that can measure the amounts of HDL and LDL/VLDL cholesterol present in plasma or serum samples within a 96-well microtiter plate format. The kit provides reagents for separating and quantifying HDL and LDL/VLDL cholesterol. The assay will detect total cholesterol (cholesteryl esters plus free cholesterol) in the presence of cholesterol esterase or only free cholesterol in the absence of the esterase enzyme. Each kit provides sufficient reagents to perform up to 192 assays, including blanks, cholesterol standards and unknown samples. Sample cholesterol concentrations are determined by comparison with a known cholesterol standard. Cholesteryl esters can be quantified by subtracting the free cholesterol values from the total cholesterol value.
Catalog No. 50-673-578 Supplier Cell Biolabs Inc Supplier No. STA391
Cell Biolabs’ HDL and LDL/VLDL Cholesterol Assay Kit is a simple fluorometric assay that can measure the amounts of HDL and LDL/VLDL cholesterol present in plasma or serum samples within a 96-well microtiter plate format. The kit provides reagents for separating and quantifying HDL and LDL/VLDL cholesterol. The assay will detect total cholesterol (cholesteryl esters plus free cholesterol) in the presence of cholesterol esterase or only free cholesterol in the absence of the esterase enzyme. Each kit provides sufficient reagents to perform up to 192 assays, including blanks, cholesterol standards and unknown samples. Sample cholesterol concentrations are determined by comparison with a known cholesterol standard. Cholesteryl esters can be quantified by subtracting the free cholesterol values from the total cholesterol value.