Learn More
Crystal Chem Inc HDL-Cholesterol Assay Kit, 96-tests
SDP
Catalog No. 501947662
Encompass_Preferred
The HDL Cholesterol Assay is used to quantify levels of HDL cholesterol in human serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 1.1 mg/dL. Dynamic range of 1.5 - 180 mg/dL
Catalog No. 50-194-7662 Supplier Crystal Chem Inc Supplier No. 80059
The HDL Cholesterol Assay is used to quantify levels of HDL cholesterol in human serum and plasma using only a 3 µL sample volume. Highlights: Kit uses sample size of only 3 µL. High sensitivity of 1.1 mg/dL. Dynamic range of 1.5 - 180 mg/dL