Learn More
Athens Research And Technology Inc HDL LIPOPROTEINS HU 5MG
Supplier:  Athens Research And Technology Inc 12160804125MG
Encompass_Preferred
Lipoproteins, High Density, Human Plasma: 5 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 175,000-360,000. Native; Purified from fresh, non-frozen human plasma in the Athens Research & Technology laboratory. Molecular Biology Reagent, Lipoproteins
Catalog No. 50-489-857