Learn More
Promocell GmbH PromoCell HDLEC-c 500,000 Human Dermal Lymphatic Endothelial Cells, adult, cryopreserved
Supplier:  Promocell GmbH C12217
Encompass
Primary Human Dermal Lymphatic Endothelial Cells (HDLEC) are a subpopulation of the Human Dermal Endothelial Cells. They are isolated from the dermis of juvenile foreskin and adult skin (different locations) from a single donor.The cells are routinely analyzed by flow cytometric analysis and immunofluorescent staining:> 95% of the cells are CD31 positive and podoplanin positive.Lymphatic vessels transport excess fluids from tissues to the circulatory system and are a major component of the immune system. The transported fluid is nearly cell free. The vessels are highly permeable, because they lack a continuous basal membrane. Lymphatic endothelial cells are involved in pathological alterations of the lymphatic system. During tumor lymphangiogenesis, the lymphatic endothelial cells build new vessels that infiltrate tumors, attract tumor cells, and induce tumor cell metastasis.
Catalog No. 50-305-972