Learn More
Abcam HDM-201 (Siremadlin), 5MG
Catalog No. 502671211
Encompass_Preferred
MW 555.4 Da, Purity >=98%. Potent and highly specific MDM-2/p53 inhibitor. It binds to human MDM2 protein with a picomolar Ki value (0.21 nM). It activates p53 and induces robust p53-dependent cell cycle arrest and apoptosis in human p53 wild-type tumor cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1211 Supplier Abcam Supplier No. AB2870605MG
MW 555.4 Da, Purity >=98%. Potent and highly specific MDM-2/p53 inhibitor. It binds to human MDM2 protein with a picomolar Ki value (0.21 nM). It activates p53 and induces robust p53-dependent cell cycle arrest and apoptosis in human p53 wild-type tumor cells.
The product is subject to the following: Abcam Restricted Use Statement