Learn More
Chondrex Inc HDM EXTRACT, 0.6 MG
Catalog No. 501526762
Encompass_Preferred
0.6 mg (lyophilized) of dust mite extract derived from Dermatophagoides pteronyssinus. Intended for use as allergen in mouse asthma models. Contact Chondrex, Inc. at suuport@chondrex.com for more information on Mouse House Dust Mite allergy model.
Catalog No. 50-152-6762 Supplier Chondrex Inc Supplier No. 3033
0.6 mg (lyophilized) of dust mite extract derived from Dermatophagoides pteronyssinus. Intended for use as allergen in mouse asthma models. Contact Chondrex, Inc. at suuport@chondrex.com for more information on Mouse House Dust Mite allergy model.