Learn More
Echelon Biosciences Research Labs 1-Hydroxy-2-methyl-2-buten-4-yl 4-diphosphate (HDMAPP) 10 mg
SDP
Catalog No. 50720406
Encompass
1-Hydroxy-2-methyl-2-buten-4-yl 4-diphosphate (HDMAPP or HMBPP) is an intermediate in the non-mevalonate pathway and is biosynthesized from 2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) by IspG (GcpE). It is the substrate for IspH (LytB) yielding IPP and DMAPP. In addition, HDMAPP is the most potent VgVd T-cell activator identified. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-406 Supplier Echelon Biosciences Research Labs Supplier No. IM05510MG
1-Hydroxy-2-methyl-2-buten-4-yl 4-diphosphate (HDMAPP or HMBPP) is an intermediate in the non-mevalonate pathway and is biosynthesized from 2-C-Methyl-D-erythritol 2,4-cyclophosphate (cMEPP) by IspG (GcpE). It is the substrate for IspH (LytB) yielding IPP and DMAPP. In addition, HDMAPP is the most potent VgVd T-cell activator identified. This item is non returnable