Learn More
Cayman Chemical HDMAPPamonIum salt 10mg
SDP
Catalog No. 502953687
Encompass_Preferred
A microbial metabolite that binds the TCR on Vγ9Vδ2 peripheral blood lymphocytes with high affinity (EC50 = 0.39 nM); induces the expansion of human memory Vγ9Vδ2 T cells
Catalog No. 50-295-3687 Supplier Cayman Chemical Supplier No. 1358010 MG
A microbial metabolite that binds the TCR on Vγ9Vδ2 peripheral blood lymphocytes with high affinity (EC50 = 0.39 nM); induces the expansion of human memory Vγ9Vδ2 T cells