Learn More
Promocell GmbH PromoCell HDMEC Pellet from > 1 million Normal Human Dermal Microvascular Endothelial Cells, pre-screened
Catalog No. 50306112
Encompass
Cell pellet consisting of 1 million cells dissolved in 200µl RNAlater for subsequent RNA, DNA or protein analysis. Note: Cell pellets cannot be revived.Our Cell Pellets are stored in RNAlater – an aqueous protection solution that rapidly permeates the cells and stabilizes cellular DNA, RNA, and proteins and protects them from degradation. This makes our Cell Pellets very useful for a wide range of research applications like gene expression analysis, cell characterization studies, and protein characterization experiments.Each Cell Pellet is made from 1 million cells and is dissolved in 200 µl RNAlater. The Cell Pellets are produced from released cell lots and have the same characteristics, i.e. expression profile or cell type specific antigens. On request, viable cells from the same lot are available for cultivation.
Catalog No. 50-306-112 Supplier Promocell GmbH Supplier No. C14017
Cell pellet consisting of 1 million cells dissolved in 200µl RNAlater for subsequent RNA, DNA or protein analysis. Note: Cell pellets cannot be revived.Our Cell Pellets are stored in RNAlater – an aqueous protection solution that rapidly permeates the cells and stabilizes cellular DNA, RNA, and proteins and protects them from degradation. This makes our Cell Pellets very useful for a wide range of research applications like gene expression analysis, cell characterization studies, and protein characterization experiments.Each Cell Pellet is made from 1 million cells and is dissolved in 200 µl RNAlater. The Cell Pellets are produced from released cell lots and have the same characteristics, i.e. expression profile or cell type specific antigens. On request, viable cells from the same lot are available for cultivation.