Learn More
US Plastic Corporation 33/400 White Polypropylene Ribbed Cap with F217 Liner
SDP
Catalog No. NC1526260
Encompass
These caps are made of polypropylene and come with a polyethylene foam liner. They are known for good impact strength and pliability, and they are cost effective. Great for shampoo, lotion, and other hair care products.
Catalog No. NC1526260 Supplier US Plastic Corporation Supplier No. 66508
These caps are made of polypropylene and come with a polyethylene foam liner. They are known for good impact strength and pliability, and they are cost effective. Great for shampoo, lotion, and other hair care products.