Learn More
Qorpak™ HDPE F-Style Jugs — Without Cap, White
Description
- Rectangular shape
- Provide moisture barrier
- Impact resistant
- For high-speed filling, pharmaceutical uses and rough handling during shipping
- Available in volumes ranging from 16 to 128 oz. (480 to 3785 mL)
- Also available with white polypropylene StudeeSeal PE foam lined cap (Cat. No. 02-991-663)
Specifications
Specifications
| Capacity (Metric) | 3840 mL |
| CAS | 67-56-1, 133454-47-4 |
| Color | White |
| Linear Formula | C24H27FN2O4 |
| MDL Number | MFCD00866688 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| No. per Case | 30/Cs. |
| Concentration | 1.0 mg/mL in methanol |
| Quantity | 30/Cs., 1 mL |
| Formula Weight | 426.48 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.