Learn More
Quality Environmental Containers HDPE Oblong Bottle, Wide Mouth, Pre-Cleaned 34 oz
SDP
Catalog No. NC2383396
Encompass_Preferred
The HDPE Oblong Wide-Mouth Bottle is designed for reliable containment in scientific applications. With a capacity of 34 oz (1 L) and a space-saving rectangular shape, it provides exceptional chemical resistance and sealing properties. Ideal for laboratory use and sensitive field sampling, it meets EPA standards with a Level 3 pre-cleaning certification. This sturdy and impact-resistant bottle is equipped with a polypropylene cap and foam liner, making it perfect for transporting and storing liquids, solids, and semi-solids.
Catalog No. NC2383396 Supplier Quality Environmental Containers Supplier No. 32120034
The HDPE Oblong Wide-Mouth Bottle is designed for reliable containment in scientific applications. With a capacity of 34 oz (1 L) and a space-saving rectangular shape, it provides exceptional chemical resistance and sealing properties. Ideal for laboratory use and sensitive field sampling, it meets EPA standards with a Level 3 pre-cleaning certification. This sturdy and impact-resistant bottle is equipped with a polypropylene cap and foam liner, making it perfect for transporting and storing liquids, solids, and semi-solids.