Learn More
Cole-Parmer Dynalon 72" Long HDPE Stirring Paddle, 3" x 6" Blade; 1/cs
Catalog No. NC1777932 Shop All Cole-Parmer Products
Encompass_Preferred
Larger size paddles and scrapers for mixing or taking samples from large tanks or vats. Stirring paddles are made of high-density polyethylene (HDPE). Nylon scraping paddles will not scratch glass.
Catalog No. NC1777932 Supplier Cole-Parmer Supplier No. 0626674
Larger size paddles and scrapers for mixing or taking samples from large tanks or vats. Stirring paddles are made of high-density polyethylene (HDPE). Nylon scraping paddles will not scratch glass.