Learn More
AVANTOR MASTERFLEX (VWR LLC) High-Density Polyethylene (HDPE) Tubing, 1/2 x 5/8"", 100 ft/pack
Catalog No. NC1463640
Encompass
Applications: General-purpose lab and industrial use. Characteristics: Low moisture absorption, good chemical and corrosion-resistance, high tensile strength. Certifications: None. Temperature range: ¿103 to 151¿F (¿75 to 66¿C) Sterilize: Autoclave.
Catalog No. NC1463640 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9550112
Applications: General-purpose lab and industrial use. Characteristics: Low moisture absorption, good chemical and corrosion-resistance, high tensile strength. Certifications: None. Temperature range: ¿103 to 151¿F (¿75 to 66¿C) Sterilize: Autoclave.