Learn More
Integrated DNA Technologies Inc Alt-R™ HDR Enhancer V2, Increases HDR Efficiency, Lowers Cytotoxicity, 150 µL
Catalog No. NC2242274
Encompass
The Alt-R™ HDR Enhancer V2 significantly boosts homology-directed repair (HDR) efficiency in genome editing workflows, providing improved cell viability with lower cytotoxicity compared to previous versions. Its versatility allows use with various delivery methods like electroporation and lipofection, making it ideal for multiple cultured human cell lines. This product uniquely combines effectiveness across diverse applications while minimizing off-target effects.
This item is not returnable. View return policy
Catalog No. NC2242274 Supplier Integrated DNA Technologies Inc Supplier No. 10007921
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.
The Alt-R™ HDR Enhancer V2 significantly boosts homology-directed repair (HDR) efficiency in genome editing workflows, providing improved cell viability with lower cytotoxicity compared to previous versions. Its versatility allows use with various delivery methods like electroporation and lipofection, making it ideal for multiple cultured human cell lines. This product uniquely combines effectiveness across diverse applications while minimizing off-target effects.