Learn More
Aviva Systems Biology Corp HDX Antibody - N-terminal region (ARP39832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HDX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HDX. Alternate names: CXorf43, D030011N01Rik, FLJ30678, MGC126769, MGC126771ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57193