Learn More
Waters Corp HDX Back Pressure Regulator Kit, 25 μL/min to 400 μL/min
Catalog No. 502391546 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Back-Pressure Regulator (BPR) is an optional attachment that applies pressure to the Enzymate digestion column. It is installed downstream of the column, specifically at the bulkhead fitting for the waste line. The blue knob on the BPR allows for manual adjustment of the back pressure. Pressure readings can be monitored in the console via the auxiliary solvent manager (ASM).
  • Back pressure regulator (BPR) for HDX
  • Mounting bracket for automated HDX systems
  • Mounting bracket for manual injection HDX systems
  • Slotted holder for BPR
  • M4 × 10 flat-head screws (2)
  • M4 × 10 blue panel screws (3)
  • Stainless steel tube assembly
  • Flow rate range: 25 μL/min to 400 μL/min
  • Regulated back pressure: Setpoint ±13,790 kPa (137.9 bar, 2,000 psi)
  • Temperature range: 15 °C to 28 °C
Catalog No. 50-239-1546 Supplier Waters Corp Supplier No. 205000969
Add to Cart
Add to Cart
Request a Quote
The Back-Pressure Regulator (BPR) is an optional attachment that applies pressure to the Enzymate digestion column. It is installed downstream of the column, specifically at the bulkhead fitting for the waste line. The blue knob on the BPR allows for manual adjustment of the back pressure. Pressure readings can be monitored in the console via the auxiliary solvent manager (ASM).
  • Back pressure regulator (BPR) for HDX
  • Mounting bracket for automated HDX systems
  • Mounting bracket for manual injection HDX systems
  • Slotted holder for BPR
  • M4 × 10 flat-head screws (2)
  • M4 × 10 blue panel screws (3)
  • Stainless steel tube assembly
  • Flow rate range: 25 μL/min to 400 μL/min
  • Regulated back pressure: Setpoint ±13,790 kPa (137.9 bar, 2,000 psi)
  • Temperature range: 15 °C to 28 °C