Learn More
Hoefer, Inc HE-FLEX Horizontal Electrophoresis Complete System
SDP
Catalog No. NC3942884
Encompass_Preferred
The HE-FLEX is completely integrated with a built-in variable power supply and provides a compact easy-to-use horizontal electrophoresis system for the separation of nucleic acids. This all-in-one system is ideal for basic academic, educational and field-testing needs.Electrophoresis TankDimensions (w x l x h) 15 x 15 x 4.0 cmMaterial Characterisitics: UV transmitting (50% at 254 nm, 80% at 312 nm)Solution Volume: 500 mlPower Supply Input: AC 100–240 V, 50/60 HzOutput: 10–150 V (10 V steps), 10–400 mA (5 mA steps), max 150 V peakMax Power: 55 WTimer:99 hours 59 min, audible alertAutomatic memory CE Safety CertifiedIncludes:Gel tank, Safety lid, Power supply, Large gel tray, 13 x 13 cm, 1 pcs, Medium gel tray, 13 x 6.5 cm, 1 pcs, Mini gel trays, 6.5 x 6.5 cm, 2 pcs, Large combs 12+12/27 wells, 1 mm thick, 4 pcs, Medium comb 9+9/19 wells, 1.5 mm thick, 1 pcs, Medium comb 7+7/14 wells, 1.5 mm thick, 1 pcs, Mini comb 2+3/3+3 wells, 2 mm thick, 1 pcs, Casting stand
Catalog No. NC3942884 Supplier Hoefer, Inc Supplier No. HEFLEX
The HE-FLEX is completely integrated with a built-in variable power supply and provides a compact easy-to-use horizontal electrophoresis system for the separation of nucleic acids. This all-in-one system is ideal for basic academic, educational and field-testing needs.Electrophoresis TankDimensions (w x l x h) 15 x 15 x 4.0 cmMaterial Characterisitics: UV transmitting (50% at 254 nm, 80% at 312 nm)Solution Volume: 500 mlPower Supply Input: AC 100–240 V, 50/60 HzOutput: 10–150 V (10 V steps), 10–400 mA (5 mA steps), max 150 V peakMax Power: 55 WTimer:99 hours 59 min, audible alertAutomatic memory CE Safety CertifiedIncludes:Gel tank, Safety lid, Power supply, Large gel tray, 13 x 13 cm, 1 pcs, Medium gel tray, 13 x 6.5 cm, 1 pcs, Mini gel trays, 6.5 x 6.5 cm, 2 pcs, Large combs 12+12/27 wells, 1 mm thick, 4 pcs, Medium comb 9+9/19 wells, 1.5 mm thick, 1 pcs, Medium comb 7+7/14 wells, 1.5 mm thick, 1 pcs, Mini comb 2+3/3+3 wells, 2 mm thick, 1 pcs, Casting stand