Learn More
Hoefer, Inc Hoefer™ HE-PLUS Horizontal Nucleic Acid Electrophoresis System (230V)
SDP
Catalog No. NC2741110
Encompass_Preferred
Complete horizontal nucleic acid electrophoresis system designed to meet a wide range of research application requirementsThe HE-PLUS is a compact easy-to-use horizontal electrophoresis system for the separation of nucleic acids. It has a built-in variable power supply. An LED display enables easy viewing of the voltage, amperage, and time. Also featured is a 99:59 minute timer with an audible end of run alert.Features:Compact and Easy to UseBuilt-in Variable Power Supply with LED DisplayAdjustable Power Supply from10 to 400 mA or 10 to 150 VoltsBuilt-in Timer with Audible AlertIncludes:Gel tank;Safety lid; Power supply; Large gel tray; Medium gel trays, 2 pieces; Large/medium comb 14/28 wells, 4 pieces; Casting standSpecifications:Unit Dimensions (W x L x H): 24.5 x 17.0 x 6.2 cmGel Dimensions (W x L): 12.5 x 13.0 cmMaximum Sample Capacity: 112 samples (4 combs, 28 samples each)Buffer Capacity: 350 mlDistance Between Electrodes: 13.5 cm
Catalog No. NC2741110 Supplier Hoefer, Inc Supplier No. HEPLUS230V
Complete horizontal nucleic acid electrophoresis system designed to meet a wide range of research application requirementsThe HE-PLUS is a compact easy-to-use horizontal electrophoresis system for the separation of nucleic acids. It has a built-in variable power supply. An LED display enables easy viewing of the voltage, amperage, and time. Also featured is a 99:59 minute timer with an audible end of run alert.Features:Compact and Easy to UseBuilt-in Variable Power Supply with LED DisplayAdjustable Power Supply from10 to 400 mA or 10 to 150 VoltsBuilt-in Timer with Audible AlertIncludes:Gel tank;Safety lid; Power supply; Large gel tray; Medium gel trays, 2 pieces; Large/medium comb 14/28 wells, 4 pieces; Casting standSpecifications:Unit Dimensions (W x L x H): 24.5 x 17.0 x 6.2 cmGel Dimensions (W x L): 12.5 x 13.0 cmMaximum Sample Capacity: 112 samples (4 combs, 28 samples each)Buffer Capacity: 350 mlDistance Between Electrodes: 13.5 cm